Anti-PGBD4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23709
10729-038EA 527.69 USD
10729-038
Anti-PGBD4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PGBD4.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: PGBD4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR