Anti-CD38 Rat Monoclonal Antibody (Spectral Red®) [clone: 90]

Supplier: Abnova
MAB5930
10707-722EA 527.69 USD
10707-722
Anti-CD38 Rat Monoclonal Antibody (Spectral Red®) [clone: 90]
Antibodies
Rat monoclonal antibody raised against native Cd38.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 90
Conjugation: Spectral Red®
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR