Anti-COQ9 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23716
10708-814EA 527.69 USD
10708-814
Anti-COQ9 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant COQ9.

Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Western Blot: 1:250-1:500

Type: Primary
Antigen: COQ9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR