Anti-TNNI3 Goat Polyclonal Antibody

Supplier: Abnova
PAB13849
10629-010EA 1572.31 USD
10629-010
Anti-TNNI3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against Troponin I-Cardiac.

Type: Primary
Antigen: TNNI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR