Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: I3/2.3]

Supplier: Abnova
MAB5937
10735-598EA 406.15 USD
10735-598
Anti-PTPRC Rat Monoclonal Antibody (Biotin) [clone: I3/2.3]
Antibodies
Rat monoclonal antibody raised against native Ptprc.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: I3/2.3
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR