Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: I3/2.3]

Supplier: Abnova
MAB5938
10733-986EA 406.15 USD
10733-986
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: I3/2.3]
Antibodies
Rat monoclonal antibody raised against native Ptprc.

Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: I3/2.3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR