Anti-CCL3 Mouse Monoclonal Antibody [clone: 426]

Supplier: Abnova
MAB6305
10734-694EA 653.85 USD
10734-694
Anti-CCL3 Mouse Monoclonal Antibody [clone: 426]
Antibodies
Mouse monoclonal antibody raised against recombinant CCL3.

Type: Primary
Antigen: CCL3
Clonality: Monoclonal
Clone: 426
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR