Anti-CCL2 Mouse Monoclonal Antibody [clone: S382]

Supplier: Abnova
MAB6317
10734-676EA 653.85 USD
10734-676
Anti-CCL2 Mouse Monoclonal Antibody [clone: S382]
Antibodies
Mouse monoclonal antibody raised against recombinant CCL2.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: S382
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR