Anti-TGFB1 Mouse Monoclonal Antibody [clone: 2C6]

Supplier: Abnova
MAB6319
10735-688EA 653.85 USD
10735-688
Anti-TGFB1 Mouse Monoclonal Antibody [clone: 2C6]
Antibodies
Mouse monoclonal antibody raised against native TGFB1.

Recommended Dilutions: Western Blot: 50-500 ng/ml

Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR