Anti-TGFB1 Mouse Monoclonal Antibody [clone: 2E6]

Supplier: Abnova
MAB6320
10735-690EA 653.85 USD
10735-690
Anti-TGFB1 Mouse Monoclonal Antibody [clone: 2E6]
Antibodies
Mouse monoclonal antibody raised against native TGFB1.

Recommended Dilutions: Western Blot: 1-10 ng/ml

Type: Primary
Antigen: TGFB1
Clonality: Monoclonal
Clone: 2000000
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR