Anti-Hepatitis C virus core protein Mouse Monoclonal Antibody [clone: A1/3D1]

Supplier: Abnova
MAB6324
10735-484EA 653.85 USD
10735-484
Anti-Hepatitis C virus core protein Mouse Monoclonal Antibody [clone: A1/3D1]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of hepatitis C virus core protein.

Type: Primary
Antigen: Hepatitis C virus core protein
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: A1/3D1
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR