Anti-NS3 Mouse Monoclonal Antibody [clone: 70-13]

Supplier: Abnova
MAB6327
10735-082EA 653.85 USD
10735-082
Anti-NS3 Mouse Monoclonal Antibody [clone: 70-13]
Antibodies
Mouse monoclonal antibody raised against partial recombinant hepatitis C virus NS-3.

Recommended Dilutions: Western Blot: 4 ?g/ml

Type: Primary
Antigen: NS3
Clonality: Monoclonal
Clone: 70-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR