Anti-APOA1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13881
10626-584EA 406.15 USD
10626-584
Anti-APOA1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Apo AI.

Type: Primary
Antigen: APOA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Hamster, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR