Anti-immunodeficiency virus type 1 P24 (core Mouse Monoclonal Antibody [clone: 163]

Supplier: Abnova
MAB6347
10735-124EA 653.85 USD
10735-124
Anti-immunodeficiency virus type 1 P24 (core Mouse Monoclonal Antibody [clone: 163]
Antibodies
Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 P24 (core protein).

Type: Primary
Antigen: immunodeficiency virus type 1 P24 (core
Clonality: Monoclonal
Clone: 163
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR