Anti-APOB Goat Polyclonal Antibody

Supplier: Abnova
PAB13887
10626-608EA 1178.46 USD
10626-608
Anti-APOB Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against APOB.

Recommended Dilutions: ELISA: 1:10000-1:80000; Western Blot: 1:200-1:10000

Type: Primary
Antigen: APOB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR