Anti-immunodeficiency virus type 1 P24 (core Mouse Monoclonal Antibody [clone: 340]

Supplier: Abnova
MAB6348
10735-494EA 653.85 USD
10735-494
Anti-immunodeficiency virus type 1 P24 (core Mouse Monoclonal Antibody [clone: 340]
Antibodies
Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 P24 (core protein).

Recommended Dilutions: Sandwich ELISA: Capture coating dose: 4 ?g/ml; Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: immunodeficiency virus type 1 P24 (core
Clonality: Monoclonal
Clone: 340
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR