Anti-NS4 Mouse Monoclonal Antibody [clone: 101]

Supplier: Abnova
MAB6342
10735-084EA 653.85 USD
10735-084
Anti-NS4 Mouse Monoclonal Antibody [clone: 101]
Antibodies
Mouse monoclonal antibody raised against recombinant hepatitis C virus NS-4.

Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: NS4
Clonality: Monoclonal
Clone: 101
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR