Anti-Chlamydia trachomatis (Elementary Bodies) Goat Polyclonal Antibody

Supplier: Abnova
PAB13905
10626-692EA 1046.15 USD
10626-692
Anti-Chlamydia trachomatis (Elementary Bodies) Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against Chlamydia trachomatis (Elementary Bodies).

Type: Primary
Antigen: Chlamydia trachomatis (Elementary Bodies)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR