Anti-Chlamydia trachomatis (Elementary Bodies) Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13906
10626-694EA 1178.46 USD
10626-694
Anti-Chlamydia trachomatis (Elementary Bodies) Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against Chlamydia trachomatis (Elementary Bodies).

Type: Primary
Antigen: Chlamydia trachomatis (Elementary Bodies)
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR