Anti-Semenogelin 1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB14672
10686-072EA 406.15 USD
10686-072
Anti-Semenogelin 1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against native Semg1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:1000-1:5000; Immunoprecipitation: 1:50-1:200; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Semenogelin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR