Anti-DIABLO Rabbit Polyclonal Antibody

Supplier: Abnova
PAB14673
10683-790EA 406.15 USD
10683-790
Anti-DIABLO Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant DIABLO.

Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:1000-1:5000; Immunoprecipitation: 1:50-1:200

Type: Primary
Antigen: DIABLO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gerbil, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR