Anti-immunodeficiency virus P24 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB14679
10627-656EA 426.15 USD
10627-656
Anti-immunodeficiency virus P24 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant human Immunodeficiency Virus (HIV) P24.

Type: Primary
Antigen: immunodeficiency virus P24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Viruses
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR