Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]

Supplier: Abnova
MAB8043
10683-754EA 920 USD
10683-754
Anti-CYCS Mouse Monoclonal Antibody [clone: 7H8.2C12]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of CYCS.

Recommended Dilutions: Western Blot: 1:500-1:1000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: 7H8.2C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR