Anti-Melatonin Rabbit Polyclonal Antibody

Supplier: Abnova
PAB14700
10725-318EA 825 USD
10725-318
Anti-Melatonin Rabbit Polyclonal Antibody
Rabbit polyclonal antibody raised against Melatonin.

Recommended Dilutions: Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Melatonin
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR