Anti-Troponin I Type 3 Mouse Monoclonal Antibody [clone: 2Q1100]

Supplier: Abnova
MAB8049
10692-172EA 1178.46 USD
10692-172
Anti-Troponin I Type 3 Mouse Monoclonal Antibody [clone: 2Q1100]
Antibodies
Mouse monoclonal antibody raised against TNNI3.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5-5 ?g/ml

Type: Primary
Antigen: Troponin I Type 3
Clonality: Monoclonal
Clone: 2Q1100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Bovine, Chicken, Frog, Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR