Anti-CXCR2 Mouse Monoclonal Antibody [clone: 6D499]

Supplier: Abnova
MAB8050
10683-718EA 1178.46 USD
10683-718
Anti-CXCR2 Mouse Monoclonal Antibody [clone: 6D499]
Antibodies
Mouse monoclonal antibody raised against native CXCR2.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5-5 ?g/ml

Type: Primary
Antigen: CXCR2
Clonality: Monoclonal
Clone: 6D499
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR