Anti-SLC2A2 Mouse Monoclonal Antibody [clone: 5D1]

Supplier: Abnova
MAB8054
10686-116EA 1178.46 USD
10686-116
Anti-SLC2A2 Mouse Monoclonal Antibody [clone: 5D1]
Antibodies
Mouse monoclonal antibody raised against SLC2A2.

Recommended Dilutions: Immunocytochemistry: 10 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 10 ?g/ml

Type: Primary
Antigen: SLC2A2
Clonality: Monoclonal
Clone: 5D1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR