Anti-Taurine Polyclonal Antibody

PAB14707
10740-556EA 910.77 USD
10740-556
Anti-Taurine Polyclonal Antibody
Rabbit polyclonal antibody raised against Taurine.

Recommended Dilutions: ELISA: 1:2000-1:10000; Immunohistochemistry: 1:1000-1:5000

Type: Primary
Antigen: Taurine
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR