Anti-COG8 Goat Polyclonal Antibody

Supplier: Abnova
PAB15620
10708-592EA 527.69 USD
10708-592
Anti-COG8 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of COG8.

Recommended Dilutions: ELISA: 1:16000

Type: Primary
Antigen: COG8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR