Anti-E2F8 Goat Polyclonal Antibody

Supplier: Abnova
PAB15624
10711-562EA 527.69 USD
10711-562
Anti-E2F8 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of E2F8.

Recommended Dilutions: ELISA: 1:32000

Type: Primary
Antigen: E2F8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR