Anti-FRMPD1 Goat Polyclonal Antibody

Supplier: Abnova
PAB15629
10715-246EA 406.15 USD
10715-246
Anti-FRMPD1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of FRMPD1.

Recommended Dilutions: ELISA: 1:2000

Type: Primary
Antigen: FRMPD1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope: internal
Host:
Isotype: Goat
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR