Anti-PARP4 Monoclonal Antibody [clone: P193-6]

Supplier: Abnova
MAB5190
10628-454EA 653.85 USD
10628-454
Anti-PARP4 Monoclonal Antibody [clone: P193-6]
Antibodies
Mouse monoclonal antibody raised against partial recombinant PARP4.

Recommended Dilutions: Immunohistochemistry (Frozen sections): 1:10-1:20

Type: Primary
Antigen: PARP4
Clonality: Monoclonal
Clone: P193-6
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR