Anti-KCNJ1 Goat Polyclonal Antibody

Supplier: Abnova
PAB15638
10721-954EA 527.69 USD
10721-954
Anti-KCNJ1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of KCNJ1.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: KCNJ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR