Anti-KCNQ3 Goat Polyclonal Antibody

Supplier: Abnova
PAB15640
10722-036EA 406.15 USD
10722-036
Anti-KCNQ3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of KCNQ3.

Recommended Dilutions: ELISA: 1:16000

Type: Primary
Antigen: KCNQ3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR