Anti-KLK1B27 Goat Polyclonal Antibody

Supplier: Abnova
PAB15642
10722-716EA 406.15 USD
10722-716
Anti-KLK1B27 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of Klk1b27.

Recommended Dilutions: ELISA: 1:1000

Type: Primary
Antigen: KLK1B27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR