Anti-MC5R Goat Polyclonal Antibody

Supplier: Abnova
PAB15646
10725-100EA 527.69 USD
10725-100
Anti-MC5R Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of MC5R.

Recommended Dilutions: ELISA: 1:8000; Western Blot: 0.3-1 ?g/ml

Type: Primary
Antigen: MC5R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR