Anti-Mcpt4 Goat Polyclonal Antibody

Supplier: Abnova
PAB15647
10725-152EA 527.69 USD
10725-152
Anti-Mcpt4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised aganist synthetic peptide of Mcpt4.

Recommended Dilutions: ELISA: 1:64000

Type: Primary
Antigen: Mcpt4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope: internal
Host:
Isotype: Goat
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR