Anti-GFAP Mouse Monoclonal Antibody [clone: GFA-02]

Supplier: Abnova
MAB5206
10715-774EA 527.69 USD
10715-774
Anti-GFAP Mouse Monoclonal Antibody [clone: GFA-02]
Antibodies
Mouse monoclonal antibody raised against native GFAP.

Type: Primary
Antigen: GFAP
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: GFA-02
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR