Anti-ANAPC10 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB19239
10688-992EA 527.69 USD
10688-992
Anti-ANAPC10 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody rasied against synthetic peptide of ANAPC10.

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunohistochemistry: 5 ?g/ml

Type: Primary
Antigen: ANAPC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR