Anti-HSP90AB1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB5725
10674-724EA 426.15 USD
10674-724
Anti-HSP90AB1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of HSP90AB1.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: HSP90AB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR