Anti-BAGE2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB17625
10690-748EA 426.15 USD
10690-748
Anti-BAGE2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of BAGE2.

Recommended Dilutions: Immunohistochemistry: 1:50-1:100; ELISA: 1:40000

Type: Primary
Antigen: BAGE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR