Anti-FLVCR1 Goat Polyclonal Antibody

Supplier: Abnova
PAB6710
10714-932EA 527.69 USD
10714-932
Anti-FLVCR1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of FLVCR1.

Recommended Dilutions: ELISA: 1:128000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: FLVCR1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR