Anti-APOD Rabbit Polyclonal Antibody

Supplier: Abnova
PAB5870
10689-672EA 406.15 USD
10689-672
Anti-APOD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Apod.

Type: Primary
Antigen: APOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR