Anti-NLRP11 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB19308
10733-026EA 527.69 USD
10733-026
Anti-NLRP11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody rasied against synthetic peptide of NLRP11.

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunocytochemistry: 2.5 ?g/ml

Type: Primary
Antigen: NLRP11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR