Anti-UBA2 Goat Polyclonal Antibody

Supplier: Abnova
PAB6480
10743-570EA 527.69 USD
10743-570
Anti-UBA2 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of UBA2.

Recommended Dilutions: ELISA: 1:16000

Type: Primary
Antigen: UBA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR