Anti-ARHGEF4 Goat Polyclonal Antibody

Supplier: Abnova
PAB6484
10690-086EA 527.69 USD
10690-086
Anti-ARHGEF4 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of ARHGEF4.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 2-4 ?g/ml

Type: Primary
Antigen: ARHGEF4
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host:
Isotype: Goat
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR