Anti-CHPF2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB17669
10707-912EA 426.15 USD
10707-912
Anti-CHPF2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of CHPF2.

Recommended Dilutions: Western Blot: 1:500-1:1000; ELISA: 1:40000

Type: Primary
Antigen: CHPF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR