Anti-ADAP1 Goat Polyclonal Antibody

Supplier: Abnova
PAB6491
10692-300EA 527.69 USD
10692-300
Anti-ADAP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of ADAP1.

Recommended Dilutions: ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: ADAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR