Anti-AGAP3 Goat Polyclonal Antibody

Supplier: Abnova
PAB6497
10688-418EA 527.69 USD
10688-418
Anti-AGAP3 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of AGAP3.

Recommended Dilutions: ELISA: 1:32000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml

Type: Primary
Antigen: AGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR