Anti-STXBP1 Goat Polyclonal Antibody

Supplier: Abnova
PAB6504
10740-004EA 527.69 USD
10740-004
Anti-STXBP1 Goat Polyclonal Antibody
Antibodies
Goat polyclonal antibody raised against synthetic peptide of STXBP1.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 0.1-0.3 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5-10 ?g/ml

Type: Primary
Antigen: STXBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR